Learn More
Crescent Chemical Co Inc CLP PEST SPIKE SOLN 7COMP 1ML
SDP
Supplier:  Crescent Chemical Co Inc CTCC2481
Encompass_Preferred
CLP Auxilliary Pesticide Solution - Low Concentration SOW-Pesticide Laboratory Control Spiking Solution, 7 Components g-BHC-100 ugmL, g-Chlordane-100 ugmL, Dieldrin-200 ugmL, 4,4 CAS Size - 1mL Catalog Number - CC2481
Catalog No. 50-683-49