Learn More
Medchemexpress LLC CLP257 | 1181081-71-9 | 99.3% | 307.34 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY110143100MG
Encompass_Preferred
CLP257 is a selective K+-Cl cotransporter KCC2 activator with an EC50 of 616 nM. It does not act on NKCC1, GABAA receptors, KCC1, KCC3, or KCC4. This compound helps restore impaired chloride transport in neurons where KCC2 activity is reduced. It has also been shown to alleviate hypersensitivity in rats experiencing neuropathic pain. CLP257 also influences plasmalemmal KCC2 protein turnover after translation.
  • Selective K+-Cl cotransporter KCC2 activator.
  • Restores impaired chloride transport in neurons.
  • Alleviates hypersensitivity in rats with neuropathic pain.
  • Modulates plasmalemmal KCC2 protein turnover post-translationally.
Catalog No. 50-003-43326