Learn More
Cayman Chemical CLS-MPEG 2000 10mg
SDP
Supplier:  Cayman Chemical 3464710 MG
Encompass_Preferred
A PEGylated form of cholesterol; has been used in the synthesis of liposomes for targeted drug delivery
Catalog No. 50-294-9889