Learn More
Lifeline Cell Technology CLSTN ALZRD OSTEO LIFELINE BTL
SDP
Supplier:  Lifeline Cell Technology CM0058
Encompass_Preferred
Lifeline's 2% Alizarin Red Stain is used to stain calcium deposited by osteocytes.
Catalog No. 50-311-923