Learn More
ABclonal Technology Clusterin alpha chain Rabbit pAb
SDP
Supplier:  ABclonal Technology A129131000UL
Encompass_Preferred
The protein encoded by this gene is a secreted chaperone that can under some stress conditions also be found in the cell cytosol. It has been suggested to be involved in several basic biological events such as cell death, tumor progression, and neurodegenerative disorders. Alternate splicing results in both coding and non-coding variants.
Catalog No. 50-000-98719