Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clz-8 | 678158-55-9 | 99.1% | 25 MG
SDP

Supplier:  Medchemexpress LLC HY12262725MG

Encompass_Preferred

CLZ-8 is an orally active small-molecule inhibitor of the Mcl-1-PUMA protein-protein interaction. It reduces PUMA-dependent apoptosis and antagonizes Mcl-1-mediated anti-apoptosis in cancer cells; supplied as a high-purity research reagent for biochemical and cellular studies.

  • Orally active Mcl-1-PUMA interface inhibitor.
  • Reported Ki of 0.3 μM for the target interaction.
  • High purity (99.1%).
  • Available in multiple milligram quantities, including 25 mg vials.
  • Molecular weight 393.50 g/mol; formula C22H23N3O2S.
  • Store powder at -20°C for long-term stability.

Catalog No. 50-002-30300


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.