Learn More
Medchemexpress LLC Clz-8 (compound 8) | 678158-55-9 | 99.1% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12262750MG
Encompass_Preferred
CLZ-8 is an orally active Mcl-1-PUMA interface inhibitor (Compound 8) used in preclinical research to modulate apoptosis; reported Ki = 0.3 μM and shows protective and anti-radiation effects in cell and animal studies.
  • Orally active Mcl-1-PUMA interface inhibitor with reported Ki = 0.3 μM.
  • Reduces PUMA-dependent apoptosis and deactivates Mcl-1-mediated anti-apoptosis.
  • High-purity solid suitable for biochemical and cellular assays.
  • Available in small research packs for laboratory use.
  • Recommended storage: powder at -20°C for long-term stability.
Catalog No. 50-002-30930