Learn More
Medchemexpress LLC HY-101925 5mg Medchemexpress, CM-272 CAS:1846570-31-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1019255MG
Encompass_Preferred
Medchemexpress, HY-101925 5mg CM-272 CAS:1846570-31-7 CM-272 is a first-in-class, potent, selective, substrate-competitive and reversible dual G9a/DNA methyltransferases (DNMTs) inhibitor. CM-272 inhibits G9a, DNMT1, DNMT3A, DNMT3B and GLP with IC50s of 8 nM, 382 nM, 85 nM, 1200 nM and 2 nM, respectively. CM-272 inhibits cell proliferation and promotes apoptosis, inducing IFN-stimulated genes and immunogenic cell death. Anti-tumour Activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9506