Learn More
TARGETMOL CHEMICALS INC CM-675 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T108415MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CM-675 is a dual inhibitor of phosphodiesterase 5 (PDE5) and class I histone deacetylases (IC50s 114 nM and 673 nM for PDE5 and HDAC1) with the potential to treat Alzheimer’s disease. purity: 99%
Catalog No. 50-258-1645