Learn More
Teknova CM Glucose Plates minus Tryptophan. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Supplier:  Teknova C3060
Encompass
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.
Catalog No. 50-238-8147