Learn More
TARGETMOL CHEMICALS INC CMF019 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T108435MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CMF019 is a potent agonist of the Apelin receptor (APJ) with G protein bias. CMF019 binds to APJ (pKis 8.58 8.49 and 8.71 for the human rat and mouse). purity: 98%
Catalog No. 50-258-1644