Learn More
Medchemexpress LLC HY-103080 5mg , CMF019 CAS:1586787-08-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1030805MG
Encompass_Preferred
HY-103080 5mg CMF019 CAS: 1586787-08-7 CMF019 is a potent and small molecule agonist at Apelin receptor (APJ) with G protein bias. CMF019 binds to APJ with pKi values of 8.58, 8.49 and 8.71 for human, rat, and mouse, respectively. CMF019 mimics the beneficial cardiovascular actions of apelin in rodents. Apelin receptor (APJ) is a G protein-coupled receptor (GPCR) activated by the endogenous peptide apelin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1623