Learn More
Medchemexpress LLC HY-52101 25mg , CMK CAS:821794-90-5 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY5210125MG
Encompass_Preferred
Medchemexpress, HY-52101 25mg CMK CAS:821794-90-5 Formula:C18H19ClN4O2 Purity:98% CMK is a RSK2 kinase inhibitor, used for cancer treatment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3994
 Restricted Item