Learn More
ATCC ATCC CMMT (CRL-6299) Rhesus Macaque Mammary Epithelial Cell Line, Frozen
Supplier: ATCC CRL6299
CMMT is a rhesus macaque (Macaca mulatta) mammary gland epithelial cell line supplied frozen for adherent culture. It is intended for laboratory research use only and requires biosafety level 2 handling. Thaw and culture in DMEM with 10% fetal bovine serum at 37°C in 5% CO2; cryopreserve in complete medium with 5% DMSO and store in vapor-phase liquid nitrogen.
- Species: Macaca mulatta (rhesus macaque).
- Tissue of origin: mammary gland (breast).
- Cell type and morphology: epithelial, adherent cells.
- Growth conditions: DMEM with 10% fetal bovine serum, 37°C, 5% CO2.
- Product format: frozen vial for culture initiation.
- Biosafety level: BSL-2; for research use only.
- Cryopreservation: freeze in complete medium with 5% DMSO; store in vapor-phase liquid nitrogen or below -130°C.
- Karyotype: modal number 64 (range 31-132).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.