Learn More
Medchemexpress LLC CMPD-39 | 2242582-40-5 | MFCD34567804 | >99.7% | C26H28FN3O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14165910MM/1ML
Encompass_Preferred
CMPD-39 is a highly selective, non-covalent inhibitor of the ubiquitin-specific protease USP30, with an IC50 of approximately 20 nM. It shows significant selectivity over other deubiquitinase family members. By inhibiting USP30's deubiquitinating activity, CMPD-39 increases ubiquitination of mitochondrial proteins (TOMM20 and SYNJ2BP), promoting phosphoubiquitin accumulation. This accelerates both mitochondrial autophagy (mitophagy) and peroxisomal autophagy (pexophagy).
  • Selective non-covalent inhibitor of USP30
  • Enhances ubiquitination of TOMM20 and SYNJ2BP
  • Promotes mitochondrial and peroxisomal autophagy
  • Restores impaired mitochondrial function in Parkinson's disease models
  • Applicable for studying USP30, autophagy pathways, and neurodegenerative disease therapies
Catalog No. 50-003-53664