Learn More
Gentarget CMV-TetR (Bsd) Lentiviral particles in PBS, 200ul/per vial, at 1 x 108 IFU/ml in PBS
Supplier:  Gentarget LVP017BSDPBS
Encompass
Pre-made tetracycline regulator (TetR) lentiviral particles expressing sequence verified tetracycline regulator gene under suCMV promoter (providing the highest TetR expression level). A blasticidin antibiotic marker under RSV promoter, allows to select transduced cells for long term expression.
Catalog No. NC3072716