Learn More
Medchemexpress LLC CN128 hydrochloride | 1335282-05-7 | 98.2% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY13106010MM/1ML
Encompass_Preferred
CN128 hydrochloride is an orally active and selective iron chelator. It is used for the research of β-thalassemia.
  • Orally active and selective iron chelator
  • High selectivity for iron(III) over other metals
  • Lacks genetic toxicity and is well-tolerated
  • Exhibits good oral bioavailability
  • Demonstrates good iron scavenging efficacy in rats
  • Iron removal efficacies are dose-dependent
Catalog No. 50-003-41149