Learn More
Cayman Chemical -CnabIchromen 10mg
SDP
Supplier:  Cayman Chemical 2625210 MG
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid; induces catalepsy, hypothermia, hypolocomotion, and antinociception in the mouse tetrad assay for cannabimimetic activity; intended for research and forensic applications
Catalog No. 50-293-4068