Learn More
Medchemexpress LLC Cndac hydrochloride | 134665-72-8 | 98.01% | 288.69 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16445B100MG
Encompass_Preferred
CNDAC hydrochloride is a metabolite of the orally active agent Sapacitabine (HY-16445), and a nucleoside analog. It induces DNA damage and apoptosis.
  • Metabolite of Sapacitabine (HY-16445)
  • Nucleoside analog
  • Induces DNA damage
  • Induces apoptosis
Catalog No. 50-003-91707