Learn More
Cayman Chemical CNDAChydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 4067210 MG
Encompass_Preferred
A nucleoside derivative and an active metabolite of sapacitabine; induces chromosomal aberrations in UV41 cells lacking XPF at 1 µM; cytotoxic to BRCA1-/- UWB1.289 and BRCA2-/- PEO1 cells (IC50s = 9.9 and 10.2 nM, respectively)
Catalog No. 50-294-2696