Learn More
Medchemexpress LLC Cnidicin | 14348-21-1 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN42075MG
Encompass_Preferred
Cnidicin is a coumarin that inhibits the degranulation of mast cells and the generation of nitric oxide (NO) in RAW 264.7 cells. It shows significant inhibition of beta-hexosaminidase release from cultured RBL-2H3 cells and nitric oxide production from activated RAW264.7 cells.
  • Inhibits the release of beta-hexosaminidase from cultured RBL-2H3 cells in a dose-dependent manner (IC50 = 25 μM)
  • Inhibits nitric oxide production from activated RAW264.7 cells (IC50 = 7.5 μM)
Catalog No. 50-004-18225