Learn More
Aviva Systems Biology Corp CNPY3 Antibody - N-terminal region (ARP57944_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57944_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNPY3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CNPY3. Alternate names: CAG4A, ERDA5, PRAT4A, TNRC5ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-419