Learn More
Medchemexpress LLC Cnqx | 115066-14-3 | C9H4N4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15066
Encompass_Preferred
CNQX (FG9065) is a potent and competitive AMPA/kainate receptor antagonist. It is a competitive non-NMDA receptor antagonist. It can block the expression of fear-potentiated startle in rats.
  • IC50s of 0.3 μM and 1.5 μM for AMPA and kainate receptors, respectively
  • Competitive non-NMDA receptor antagonist
  • Blocks the expression of fear-potentiated startle in rats
Catalog No. 50-003-74469