Learn More
Medchemexpress LLC CNTF Human 100ug

Supplier: Medchemexpress LLC HYP7146100UG
Recombinant human ciliary neurotrophic factor (CNTF) supplied lyophilized for research use. CNTF is a pluripotent cytokine in the IL-6 family that promotes survival, differentiation, and neurite outgrowth of central and peripheral neurons, including retinal photoreceptors. The protein is produced in E. coli, supplied with high purity, and formulated for stable storage and convenient reconstitution.
- Supports neuronal survival and differentiation.
- Protects retinal cone and rod photoreceptors.
- Produced in E. coli and supplied tag-free.
- Purity greater than 95% by reducing SDS-PAGE.
- Lyophilized formulation for stable storage.
- Recommended storage at -20°C; aliquot and store at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.