Learn More
Sino Biological CNTF/Ciliary Neurotrophic Factor Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11841MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CNTF/Ciliary Neurotrophic Factor (rh CNTF/Ciliary Neurotrophic Factor; Catalog#11841-H07E; NP_000605.1; Ala2-Met200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8655