Learn More
Cayman Chemical CntharIdIc AcIdodIum salt 10mg
SDP
Supplier:  Cayman Chemical 1907310 MG
Encompass_Preferred
A PP1 and PP2A inhibitor with IC50 values of 0.6 and 0.05 µM, respectively
Catalog No. 50-298-2450