Learn More
Medchemexpress LLC Cnx-2006 | 1375465-09-0 | MFCD26142654 | 98.4% | C26H27F4N7O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1389710MG
Encompass_Preferred
CNX-2006 is a mutant-selective, irreversible epidermal growth factor receptor (EGFR) inhibitor with potent activity against the EGFRT790M mutant (IC50 ≈ 20 nM). It is supplied for research use as a solid and in DMSO solutions, and selectively inhibits growth of EGFR-T790M cells versus wild-type EGFR.
  • Potent mutant-selective EGFR inhibition (EGFRT790M IC50 ≈ 20 nM).
  • Irreversible mechanism of action.
  • Shows activity against uncommon EGFR mutations including G719S, L861Q, exon 19 insertion, and T854A.
  • Available as a white to gray solid and as DMSO solution for assays.
  • High purity suitable for research applications (≈98.4%).
Catalog No. 50-002-61619