Learn More
Medchemexpress LLC CO23 | 1370363-74-8 | 98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY130012
Encompass_Preferred
CO23 is a selective thyroid hormone receptor (TR) α agonist, used for growth and development regulation. It is intended for research use only and is not sold to patients. This compound has a molecular weight of 592.17 and a chemical formula of C19H18I2N2O4. It appears as an off-white to light yellow solid and is soluble in DMSO at 250 mg/mL.
  • Selective thyroid hormone receptor alpha agonist.
  • Shows EC50 values for human TRalpha transactivation activity in HeLa cells (11 nM) and U2OS cells (34 nM).
  • Demonstrates approximately 50% cholesterol reduction in hypothyroid Wistar rats at a dose of 0.8 nmol/g.
  • Increases Dio1 mRNA in hypothyroid Wistar rats at doses of 0.8, 2.5, and 5 nmol/g.
  • Soluble in DMSO at 250 mg/mL (422.18 mM) in vitro.
Catalog No. 50-003-42654