Learn More
Creative Biomart Native Human F8; Antihemophilic factor human
Supplier:  Creative Biomart THP0051
Encompass_Preferred
The product, also known as Coagulation Factor VIII or Anti-Hemophilic Factor (AHF), is a non-recombinant, lyophilized concentrate of coagulation factor VIII, an endogenous protein and essential component of the coagulation cascade. Antihemophilic factor is manufactured with reduced amounts of von Willebrand Factor antigen (VWF:Ag) and purified from extraneous plasma-derived protein by affinity chromatography. The small amount of VWF:Ag is used to purify factor VIII complex and then removed from the final preparation. The final purified concentrate contains albumin as a stabilizer.
Catalog No. 50-208-4102