Learn More
Medchemexpress LLC Coagulation factor X 10ug
SDP
Supplier:  Medchemexpress LLC HYP7423810UG
Encompass_Preferred
Coagulation factor XIII B/F13B Proteinas the B chain of factor XIII lacks catalytic activity but stabilizes A subunits and regulates thrombin-initiated transglutaminase formation Functioning as a tetramer with two A chains (F13A1) and two B chains (F13B) it structurally supports the factor XIII complex emphasizing its regulatory role in blood coagulation particularly in transglutaminase activation by thrombin Coagulation factor XIII B/F13B Protein Human (HEK293 His) is the recombinant human-derived Coagulation factor XIII B/F13B protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78794