Learn More
ACROBiosystems Cynomolgus Coagulation factor XI Protein, His Tag (SPR verified)
Supplier:  ACROBiosystems FXIC52H350UG
Encompass_Preferred
Cynomolgus Coagulation factor XI, His Tag (FXI-C52H3) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Ala 624 (Accession # G7MSF8-1).
Catalog No. 50-233-1979