Learn More
Grainger HYFLEX Coated Gloves, L, 9
Supplier:  Grainger 4JU93
Encompass_Preferred
The HyFlex 11-926 Coated Gloves are a pair of lightweight, seamless knit gloves coated with a sandy nitrile coating on the palm and fingertips for enhanced grip and dexterity. They are 9 mil thick and provide abrasion resistance. The breathable nylon liner helps keep hands cool and comfortable. The gloves are powder-free, have an elastic wrist, and are ideal for light duty applications requiring dexterity where wet or oily conditions are encountered. They meet ANSI cut level 1 standards. The gloves come in sizes S-2XL in a gray/black color.
Catalog No. NC0729341