Learn More
Strem, An Ascensus Company CAS# 3317-67-7. 1g. Cobalt(II) phthalocyanine. MFCD00010718
SDP
Supplier:  Strem, An Ascensus Company 2704881G
Encompass
CAS# 3317-67-7. 1g. Cobalt(II) phthalocyanine. MFCD00010718. Molecular Weight: 571.47. Molecular Formula: (C32H16N8)Co. Color/form: purple xtl. Strem# 27-0488. http://www.strem.com/catalog/v/27-0488/
Catalog No. 50-901-11178