Learn More
Med Vet International Cohesive Bandage 3M Coban 1 inch X 5 Yard Self-Adherent Closure Tan Nonsterile Standard Compression, 2081T, 72/Case
Supplier:  Med Vet International 1211280CS
Encompass_Preferred
This 3M Coban self-adherent wrap sticks to itself, eliminating the need for fasteners The tan, self-adhering, elastic wrap remains in place so that it doesn't need to be readjusted Compression wrap is lightweight and prevents drag on dressings, bandages and devices Its porous and breathable construction allows air to pass through to prevent the accumulation of heat This 1-inch x 5-yard elastic compression wrap won't stick to skin, dressings or bandages It stretches to provide a snug fit and accommodate contours
Catalog No. 50-283-2347
 Restricted Item