Learn More
Medchemexpress LLC HY-10493 50mg , Cobicistat CAS:1004316-88-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1049350MG
Encompass_Preferred
HY-10493 50mg Cobicistat CAS: 1004316-88-4 Cobicistat is a potent and selective inhibitor of cytochrome P450 3A (CYP3A) enzymes with IC50s of 30-285 nM. Cobicistat is a pharmacokinetic enhancer which increases the overall absorption of several HIV medications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9916