Learn More
Medchemexpress LLC HY-13064 100mg , Cobimetinib CAS:934660-93-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13064100MG
Encompass_Preferred
HY-13064 100mg Cobimetinib CAS: 934660-93-2 Cobimetinib (GDC-0973, RG7420) is a potent, selective and oral MEK1 inhibitor with an IC50 of 4.2 nM for MEK1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0700