Learn More
Apexbio Technology LLC Cobimetinib, 934660-93-2, MFCD22124461, 5 mg
SDP
Supplier:  Apexbio Technology LLC A33215
Encompass_Preferred
MF: C21H21F3IN3O2, MW: 531.31, Solubility: 26.55 mg/mL in DMSO, 33.53 mg/mL in EtOH with gentle warming,insoluble in H2O. Cobimetinib is a selective inhibitor of mitogen-activated protein kinase kinase (MEK) with IC50 value of 0.9 nM.
Catalog No. 50-115-0564