Learn More
Medchemexpress LLC HY-13064A 5mg Medchemexpress, Cobimetinib (hemifumarate) CAS:1369665-02-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13064A5MG
Encompass_Preferred
Medchemexpress, HY-13064A 5mg Cobimetinib (hemifumarate) CAS:1369665-02-0 Cobimetinib hemifumarate is a novel selective MEK1 inhibitor, and the IC 50 value against MEK1 is 4.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2653