Learn More
Cayman Chemical CoblopasvIr 10mg
SDP
Supplier:  Cayman Chemical 3677910 MG
Encompass_Preferred
A pan-genotypic inhibitor of HCV NS5A; inhibits viral replication of the HCV genotypes 1a, -1b, -3a, -4a, -5a, and -6a in cellular assays and genotype 2a in a cell-free assay
Catalog No. 50-291-0023