Learn More
Hach Company COD2 Digestion Vials, HR, Mercury-free, pk/25
Supplier:  Hach Company 2565125
Encompass
For high range, mercury-free Chemical Oxygen Demand (COD) determinations. COD2 Digestion Vials, Mercury-Free Formulation. Method: Reactor Digestion. Range: 20-1500 mg/L COD. Pack of 25 vials
Catalog No. 50-199-6241