Learn More
Hach Company COD2 Digestion Vials, LR, Mercury-free, pk/25
Supplier:  Hach Company 2565025
Encompass
For low range, mercury-free Chemical Oxygen Demand (COD) determinations. COD2 Digestion Vials, Method: Reactor Digestion Range: 0-150 mg/L COD. Pack of 25 vials.
Catalog No. 50-199-6239