Learn More
Hach Company COD Digestion Vials, Low Range, pk/25
Supplier:  Hach Company 2125825
Encompass
For determination of low range Chemical Oxygen Demand (COD) by the Reactor Digestion method. US EPA approved for wastewater analysis using Hach Method 8000. Range: 3-150 mg/L COD. Pack of 25 vials
Catalog No. 50-199-5460