Learn More
Hach Company COD in salt-/seawater - TNTplus Vial Test, HR (70 - 700 mg/L COD), 25 Tests
Supplier:  Hach Company TNT816
Encompass
For saline COD samples with Chloride concentrations up to 20,000 mg/L Cl-!COD TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. For photometric determination of high range COD by the reactor digestion method. High Chloride concentrations usually interfere COD testing and lead to a high bias. Diluting samples to reduce the Chloride concentration may lead to high variability in COD results. TNT816 is a safe, reliable and very easy to use method for the photometric detection of COD in saltwater samples, e.g. industrial wastewater. The Direct Read Technology will provide your COD result to the nearest mg/L! No more sample dilutions, cleaning glassware and dealing with large amounts of hazardous reagents and waste. Ideal to combine with our ezCOD recycling service!25 COD vials per box.
Catalog No. 50-199-8108