Learn More
Hach Company Hach Chemical Oxygen Demand (COD) TNTplus Vial Test, High Range (20-1500 mg/L COD), 150 tests (6 x 25)
Supplier:  Hach Company TNT82206PK
Encompass
The Hach Chemical Oxygen Demand (COD) TNTplus vial test kit provides a reliable, high-range method for measuring COD in wastewater and industrial samples. Pre-measured reagent vials simplify preparation: add sample, digest in a reactor block, then measure spectrophotometrically for accurate results.
  • High-range measurement: 20-1500 mg/L COD.
  • Pack contains 150 tests (6 x 25 vials).
  • Uses reactor digestion method for sample preparation.
  • Requires a reactor block for digestion and a spectrophotometer for measurement.
  • Freeze-dried, pre-measured reagents minimize handling errors.
  • Suitable for wastewater and industrial influent testing.
Catalog No. NC3244651