Learn More
Cerilliant Corporation CODEINE-D6 1ML
Supplier:  Cerilliant Corporation C041
Encompass
Codeine-D6 | 1007844-34-9 | MW 305.40 | 1mL | Codeine-D6 is a stable-labeled internal standard suitable for many LC/MS or GC/MS testing applications including pain prescription monitoring, forensic analysis, clinical toxicology, urine drug testing or isotope dilution methods. Codeine is an opiate used for its analgesic, antitussive, and sedative properties and is the second-most predominant alkaloid in opium. Since opiate drugs are classified as controlled substances, Cerilliant offers most opiate reference standards as DEA-exempt solutions for added convenience.
Catalog No. NC2326118