Learn More
Medchemexpress LLC Codlemone | 33956-49-9 | MFCD00009992 | 95.2% | 182.30 g/mol | C12H22O | 10G

Supplier: Medchemexpress LLC HY12446610G
Codlemone (codlelure) is a natural pheromone compound used in research on insect behavior and pheromone chemistry. It corresponds to (E,E)-8,10-dodecadien-1-ol (CAS 33956-49-9) with chemical formula C12H22O and molecular weight 182.30 g/mol. Typical purity is ~95.2%. The material is white to off-white and shows a temperature-dependent physical state, supplied in small laboratory quantities.
- Natural pheromone used in behavioral studies.
- High purity for analytical applications.
- White to off-white appearance.
- Temperature-dependent physical state (solid <30 °C, liquid >32 °C).
- Chemical formula C12H22O, molecular weight 182.30 g/mol.
- Supplied in small laboratory quantities (for example, 10 g).
- Provided with COA and SDS for traceability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.