Learn More
Medchemexpress LLC Coelenterazine | 55779-48-1 | 98.5% | 423.46 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY18743
Encompass_Preferred
Coelenterazine is a luminescent enzyme substrate for apoaequorin and Renilla luciferase. It has been utilized as a bioluminescence donor in bioluminescence resonance energy transfer (BRET) for detecting protein-protein interactions. It also serves as a superoxide anion-sensitive chemiluminescent probe and can be employed in the chemiluminescent detection of peroxynitrite.
  • Functions as a luminescent enzyme substrate
  • Used as a bioluminescence donor in BRET for protein-protein interaction detection
  • Acts as a superoxide anion-sensitive chemiluminescent probe
  • Can be used for chemiluminescent detection of peroxynitrite (Ex/Em = 429/466 nm)
Catalog No. 50-003-85550