Learn More
Selleck Chemical LLC Coenzyme Q1 E4447-100MG
SDP
Supplier:  Selleck Chemical LLC E4447100MG
Encompass_Preferred
Coenzyme Q1 (CoQ1) is localized to the hydrophobic domain of the phospholipid bilayer of mitochondria plasma lipoproteins and other biological membranes CoQ1 reduction has a strong rotenone-insensitive component and can be used to measure the mitochondrial respiratory chain complex 1 activity
Catalog No. 50-000-38947