Learn More
Medchemexpress LLC HY-N0111 200mg Medchemexpress, Coenzyme Q10 CAS:303-98-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0111200MG
Encompass_Preferred
Medchemexpress, HY-N0111 200mg Coenzyme Q10 CAS:303-98-0 Coenzyme Q10 is an essential cofactor of the electron transport chain and a potent antioxidant agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3401