Learn More
Cusabio Technology LLC Human Coenzyme Q10 (CoQ10) ELISA Kit
Supplier:  Cusabio Technology LLC CSBE14081H
Encompass_Preferred
This Human CoQ10 ELISA Kit was designed for the quantitative measurement of Human CoQ10 protein in serum, plasma, tissue homogenates. It is a Competitive ELISA kit, its detection range is 3.12 ng/mL-50 ng/mL and the sensitivity is 1.56 ng/mL. One kit contains 96 wells.
Catalog No. NC2952955